Learn More
Chloroiodomethane, 98%, stab. with copper
CAS: 593-71-5 | CH2ClI | 176.381 g/mol
Supplier: Thermo Scientific Chemicals A1103218
Description
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.
Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.
Notes
It is incompatible with strong oxidizing agents and strong bases.
Specifications
| Chloroiodomethane | |
| 593-71-5 | |
| 108°C to 109°C | |
| CH2ClI | |
| MFCD00001078 | |
| UN2810 | |
| Light sensitive | |
| Miscible with acetone,benzene,diethyl ether and alcohol. | |
| C(Cl)I | |
| 176.381 | |
| 176.38 |
| Stabilized with copper | |
| 2.416 | |
| None | |
| 1.5822 | |
| 50 g | |
| 1730802 | |
| chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@ | |
| PJGJQVRXEUVAFT-UHFFFAOYSA-N | |
| chloro(iodo)methane | |
| 11644 | |
| 98% |
Chemical Identifiers
| 593-71-5 | |
| MFCD00001078 | |
| chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@ | |
| chloro(iodo)methane |
| CH2ClI | |
| PJGJQVRXEUVAFT-UHFFFAOYSA-N | |
| 11644 |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 209-804-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only