missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stab. with copper

Catalog No. AAA1103209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1103209 10 g
AAA1103218 50 g
AAA1103230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1103209 Supplier Thermo Scientific Chemicals Supplier No. A1103209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 593-71-5 | CH2ClI | 176.381 g/mol

Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.

Notes
It is incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 593-71-5
Molecular Formula CH2ClI
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane

Specifications

Density 2.416
Boiling Point 108°C to 109°C
Flash Point None
Refractive Index 1.5822
Quantity 10 g
UN Number UN2810
Beilstein 1730802
Sensitivity Light sensitive
Solubility Information Miscible with acetone,benzene,diethyl ether and alcohol.
SMILES C(Cl)I
Molecular Weight (g/mol) 176.381
Formula Weight 176.38
Percent Purity 98%
Chemical Name or Material Chloroiodomethane, Stabilized with copper
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 209-804-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only