Learn More
Chlorogenic acid
An analog of caffeic acid | CAS: 327-97-9 | C16H18O9 | 354.31 g/mol
$69.01 - $603.74
Chemical Identifiers
| CAS | 327-97-9 |
|---|---|
| Molecular Formula | C16H18O9 |
| Molecular Weight (g/mol) | 354.31 |
| MDL Number | MFCD00003862 |
| InChI Key | CWVRJTMFETXNAD-JUHZACGLSA-N |
| Synonym | chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri |
| PubChem CID | 1794427 |
| ChEBI | CHEBI:16112 |
| IUPAC Name | (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
| SMILES | O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60457MD
|
Thermo Scientific Chemicals
J60457MD |
250 mg |
N/A
|
|
|||||
|
AAJ6045703
|
Thermo Scientific Chemicals
J6045703 |
1 g |
N/A
|
|
|||||
|
AAJ6045706
|
Thermo Scientific Chemicals
J6045706 |
5 g |
N/A
|
|
|||||
Description
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
Solubility
Soluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 327-97-9 | |
| 354.31 | |
| CWVRJTMFETXNAD-JUHZACGLSA-N | |
| 1794427 | |
| (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
| C16H18O9 | |
| MFCD00003862 | |
| chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
| CHEBI:16112 | |
| O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
Specifications
| 327-97-9 | |
| C16H18O9 | |
| 250 mg | |
| chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
| CWVRJTMFETXNAD-JUHZACGLSA-N | |
| (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid | |
| 1794427 | |
| 354.31 | |
| Chlorogenic acid |
| 210°C (decomposition) | |
| MFCD00003862 | |
| 14,2142 | |
| Soluble in ethanol and acetone. | |
| O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O | |
| 354.31 | |
| CHEBI:16112 | |
| Powder |
Safety and Handling
EINECSNumber : 206-325-6
RTECSNumber : GU8480000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only