missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorogenic acid, 99.45%, For HPLC analysis, MP Biomedicals™
$205.86 - $545.69
Chemical Identifiers
| CAS | 327-97-9 |
|---|---|
| Molecular Formula | C16H18O9 |
| Molecular Weight (g/mol) | 354.31 |
| MDL Number | MFCD00003862 |
| InChI Key | CWVRJTMFETXNAD-JUHZACGLSA-N |
| Synonym | chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri |
| PubChem CID | 1794427 |
| ChEBI | CHEBI:16112 |
| IUPAC Name | (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
| SMILES | O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15061801
|
MP Biomedicals Inc
0215061801 |
1 g |
Each for $205.86
|
|
|||||
|
ICN15061805
|
MP Biomedicals Inc
0215061805 |
5 g |
Each for $545.69
|
|
|||||
Chemical Identifiers
| 327-97-9 | |
| 354.31 | |
| CWVRJTMFETXNAD-JUHZACGLSA-N | |
| 1794427 | |
| (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
| C16H18O9 | |
| MFCD00003862 | |
| chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
| CHEBI:16112 | |
| O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
Specifications
| 327-97-9 | |
| 99.45% | |
| MFCD00003862 | |
| chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
| O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O | |
| 354.31 | |
| CHEBI:16112 | |
| Crystalline Powder |
| White | |
| C16H18O9 | |
| 1 g | |
| CWVRJTMFETXNAD-JUHZACGLSA-N | |
| (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid | |
| 1794427 | |
| 354.3 | |
| Chlorogenic acid, For HPLC analysis |