missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™

High purity solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

$831.69 - $831.69

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
C297-4
SDS View Documents
Thermo Fisher Scientific
BPC2974
4 L Amber Soda Lime Glass
Each for $831.69
Case of 4 Each for $3,326.76
Only null left
 
Description

Description

  • Contains approx. 50ppm amylene as preservative
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
trichloromethane
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl

Specifications

-63°C
0.01 AU max. at 280nm, 0.15 AU max. at 254nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm
1.480g/cm³
4 L
Amber Soda Lime Glass
1.4430 to 1.4460 (at 25°C)
99.9% min by GC
MFCD00000826
0.56 mPa/s at 20°C
0.01% max.
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chloroform, Approx. 50ppm Amylene as Preservative
67-66-3,513-35-9
Colorless
61°C
Pass Test
Liquid
1ppm max.
CHCl3
213mbar at 20°C
1888
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.9%
50ppm Amylene as Preservative
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id

Signal Word: WARNING!

SDS
missing translation for 'documents'

missing translation for 'documents'