missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Spectrum™ Chemical

Catalog No. p-7165286
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609690 100 mL Amber Glass Bottle
18609694 500 mL Amber Glass Bottle
18609689 1 L Amber Glass Bottle
18609693 4 L Amber Glass Bottle
18609691 20 L Amber Glass Bottle
18609692 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609690 Supplier Spectrum Chemical Mfg Cor Supplier No. C1121100ML
Only null left

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 1
Boiling Point 61°C
Quantity 100 mL
Packaging Amber Glass Bottle
Grade Ungraded
Chemical Name or Material Chloroform

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.