missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Spectrophotometric Grade, 99.5+%

Catalog No. AA32442K7
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32442K7 4 L
AA32442K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA32442K7 Supplier Thermo Scientific Chemicals Supplier No. 32442K7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

Chloroform is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point °C to 63°C
Color Colorless
Appearance Colorless liquid
Density 1.492
Quantity 4 L
Odor Characteristic
Physical Form Liquid
Refractive Index 1.446
Assay Percent Range >99.5%
Vapor Pressure 210 hPa (158mm Hg) at 20°C
Beilstein 1731042
Solubility Information Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide.
Boiling Point 60.5°C to 61.5°C
Formula Weight 119.38
Grade Spectrophotometry
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category H302-H315-H319-H331-H335-H336-H351-H361d-H373
Hazard Statement Wear suitable protective clothing and gloves.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1; Packing Group: III
EINECSNumber 200-663-8
RTECSNumber FS9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only