missing translation for 'onlineSavingsMsg'
Learn More

Chloroform Solution, NMR Reference Standard, 5% in acetone-d6 (99.9 atom % D), Chromium(III) acetylacetonate 2%, MilliporeSigma™ Supelco™

Catalog No. 111017067
Change view
Click to view available options
Quantity:
3.175 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017067 3.175 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017067 Supplier Merck Emd Millipore Supplier No. 7364308IN
Only null left

NMR Tube Size: 10 mm 8 in.

Specifications

Packaging NMR Tube Size: 10 mm 8 in.
Concentration 5% in acetone-d 6 (99.9 atom% D)
MDL Number MFCD00000826
Quantity 3.175 mL
Grade NMR Reference Standard