missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Purified, Reagents

Catalog No. 01337323
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337323 1 L
01337322 2.5 L
01337321 20 L
01337320 4 L
01337319 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337323 Supplier Labchem Inc Supplier No. C21755001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chloroform, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Trichloromethane, Methyl trichloride
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.5°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.484
Boiling Point 60.5°C to 61.2°C
Grade Pure
Chemical Name or Material Chloroform, Purified