missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Purified, Reagents
Specifications
Specifications
| Melting Point | -63.5°C |
| Color | Colorless |
| Quantity | 20 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 1.484 |
| Boiling Point | 60.5°C to 61.2°C |
| Grade | Pure |
| Chemical Name or Material | Chloroform, Purified |