missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform Molecular Biology MP Biomedicals
$138.22 - $138.22
Chemical Identifiers
| CAS | 67-66-3 |
|---|---|
| Molecular Formula | CHCl3 |
| Molecular Weight (g/mol) | 119.37 |
| MDL Number | MFCD00000826 |
| InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
| Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
| PubChem CID | 6212 |
| ChEBI | CHEBI:35255 |
| SMILES | ClC(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19400290
|
MP Biomedicals Inc
0219400290 |
0.5 L |
Each for $138.22
|
|
|||||
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| 6212 | |
| ClC(Cl)Cl |
| CHCl3 | |
| MFCD00000826 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| CHEBI:35255 |
Specifications
| 67-66-3 | |
| 0.5 L | |
| MFCD00000826 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| chloroform | |
| 6212 | |
| 119.38 | |
| Chloroform |
| 4.36 (air = 1) | |
| CHCl3 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| ClC(Cl)Cl | |
| 119.37 | |
| CHEBI:35255 | |
| Molecular Biology |