missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, MilliporeSigma™
$131.46 - $507.63
Chemical Identifiers
| CAS | 67-66-3 |
|---|---|
| Molecular Formula | CHCl3 |
| Molecular Weight (g/mol) | 119.37 |
| MDL Number | MFCD00000826 |
| InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
| Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
| PubChem CID | 6212 |
| ChEBI | CHEBI:35255 |
| IUPAC Name | trichloromethane |
| SMILES | ClC(Cl)Cl |
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| 6212 | |
| trichloromethane |
| CHCl3 | |
| MFCD00000826 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| CHEBI:35255 | |
| ClC(Cl)Cl |
Specifications
| 67-66-3 | |
| Glass Bottle | |
| CHCl3 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| ClC(Cl)Cl | |
| 119.37 | |
| CHEBI:35255 | |
| Chloroform |
| 500 mL | |
| CHCl3 | |
| MFCD00000826 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane | |
| 6212 | |
| 119.38 |