missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, MilliporeSigma™

Catalog No. MCX10601
Change view
Click to view available options
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
MCX10601 Glass Bottle
MCX10591 Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MCX10601 Supplier Technidata Supplier No. CX10601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane

Specifications

Packaging Glass Bottle
Linear Formula CHCl3
Formula Weight 119.38
Chemical Name or Material Chloroform