missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, MilliporeSigma™
Specifications
Specifications
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Linear Formula | CHCl3 |
| Formula Weight | 119.38 |
| Chemical Name or Material | Chloroform |