missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, HPLC, 99.8% min., ACS, MilliporeSigma™

Catalog No. MCX10501
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX10501 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX10501 Supplier Technidata Supplier No. CX10501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Grade ACS/HPLC/NF/USP
Physical Form Liquid
Assay Percent Range 99.8%, min.
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Chemical Name or Material Chloroform, For ACS analysis

Specifications

Quantity 4 L
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
Recommended Storage Room Temperature