missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, HPLC, 99.8%, MilliporeSigma™

Catalog No. 50947850
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle (Pre-packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50947850 4 L Glass Bottle (Pre-packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947850 Supplier Technidata Supplier No. CX10581

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point −63°C
Quantity 4 L
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 2 ppm max
Assay Percent Range 99.8% min
Vapor Pressure 211 hPa (20°C)
Boiling Point 61°C (1013 hPa)
Solubility Information 8.7 g/L
Recommended Storage Store at room temperature