missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform For HPLC, Spectrophotometry and Gas Chromatography, OmniSolv™ MilliporeSigma™
Specifications
Specifications
| Melting Point | -63°C |
| Color | Colorless |
| Quantity | 4 L |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Physical Form | Clear liquid |
| Residue after Evaporation | 1ppm |
| Titratable Acid | 0.2µeq/g |
| Vapor Pressure | 211 hPa (20°C) |
| UN Number | UN 1888 |
| Show More |
Safety and Handling
| Hazard Statement | H302 Harmful if swallowed. H315 Causes skin irritation. H319 Causes serious eye irritation. H331 Toxic if inhaled. H351 Suspected of causing cancer. H361d Suspected of damaging the unborn child. H372 Causes damage to organs (Liver, Kidney) through prolonged or repeated exposure. |
| Precautionary Statement | P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P304 + P340 IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P308 + P310 IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician. |
| EINECSNumber | 200-663-8 |