missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, For gas chromatography ECD and FID SupraSolv™ MilliporeSigma™

Catalog No. M1024322500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1024322500 2.5 L Glass Bottle
M1024321000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1024322500 Supplier Technidata Supplier No. 1.02432.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Density 1.4g/cm3 at 20°C
Quantity 2.5 L
Odor Sweet
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 211 hPa (20°C)
UN Number 1888
Boiling Point 61°C
Solubility Information Soluble in water: 8g/L at 20°C
Formula Weight 119.38
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category H302: Harmful if swallowed.
Hazard Statement H315: Causes skin irritation.
Precautionary Statement H319: Causes serious eye irritation.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.