missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, CHROMASOLV™, for HPLC, ≥99.8%, amylene stabilized, Honeywell Riedel-de Haën™

Catalog No. p-7111860
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001732 1 L Glass Bottle
6001734 2.5 L Glass Bottle
6001740 6 x 1 L Glass Bottle
6001738 4 x 2.5 L Glass Bottle
6001739 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6001732 Supplier Honeywell Chemicals Supplier No. 348541L
Only null left

CHROMASOLV™, for HPLC,≥99.8%, amylene stabilized

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C (literature)
Absorbance max. 1.00 at 250 nm, max. 0.30 at 260 nm, max. 0.005 280 nm
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.)
Boiling Point 61°C
Quantity 1 L
Packaging Glass Bottle
Linear Formula CHCl3
UN Number UN1888
Beilstein 1731042
Formula Weight 119.38
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Chloroform, CHROMASOLV™, for HPLC, ≥99.8%, amylene stabilized
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
EINECSNumber 200-663-8
Recommended Storage Room Temp