missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Anhydrous, 99% min., DriSolv, MilliporeSigma™

Catalog No. MCX10577
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX10577 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX10577 Supplier Technidata Supplier No. CX10577

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Quantity 100 mL
Physical Form Liquid
Assay Percent Range 99%, min.
Linear Formula CHCl3
Formula Weight 119.38
Chemical Name or Material Chloroform, DriSolv™
Recommended Storage Room Temperature