missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ACS, Reagents

Catalog No. 01338053
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Glass Bottle
Safety Coated Glass
Steel Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338053 1 L Safety Coated Glass
01338051 2.5 L Safety Coated Glass
01338049 20 L Steel Pail
01338048 4 L Glass Bottle
01338047 4 L Safety Coated Glass
01338046 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01338053 Supplier Labchem Inc Supplier No. C11755001C1
Only null left

Chloroform, ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Trichloromethane, Methyl trichloride
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.5°C
Color Colorless
Quantity 1 L
Packaging Safety Coated Glass
Physical Form Liquid
Specific Gravity 1.484
Boiling Point 60.5°C
Grade ACS
Chemical Name or Material Chloroform, ACS