missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ACS, Reagents
Chloroform, ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1175500200E
Specifications
| Chloroform, ACS | |
| 67-66-3 | |
| 60.5°C | |
| Steel Drum | |
| 1.484 | |
| Trichloromethane, Methyl trichloride | |
| ClC(Cl)Cl | |
| 119.37 |
| -63.5°C | |
| Colorless | |
| 200 L | |
| Liquid | |
| CHCl3 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane | |
| ACS |
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| Trichloromethane, Methyl trichloride | |
| ClC(Cl)Cl |
| CHCl3 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane |