missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ACS Reagent, With Stabilizer (Amylene), ≥99.8%, Solstice

Catalog No. 60046956
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046956 1 L Glass Bottle
60046957 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046956 Supplier Solstice Supplier No. 4724761L

ACS Reagent, contains amylenes as stabilizer,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -61.3°C
Density 1.492g/mL at 25°C, 1.48g/mL at 25°C
Boiling Point 61°C
Odor Sweet
Packaging Glass Bottle
Quantity 1 L
UN Number UN1888
Beilstein 1731042
IUPAC Name chloroform
Vapor Density 4.1 (vs air)
Formula Weight 119.38g/mol
Vapor Pressure 160mmHg (20°C)
Percent Purity ≥99.8%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Chloroform, Containsamylenes as stabilizer
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338-P311
DOTInformation 6.1/PGIII
RTECSNumber 200-663-8