missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ACS Reagent Grade, Ricca Chemical

Catalog No. RSOC00201C
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOC00201C 1 L Amber Glass Bottle
RSOC00204C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RSOC00201C Supplier Ricca Chemical Company Supplier No. RSOC00201C

For General Use

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Boiling Point 61°C
Percent Purity >99.8%
Grade ACS
Chemical Name or Material Chloroform
ShelfLife 60 months