missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99+%, for spectroscopy, stabilized with amylene, Thermo Scientific™

Catalog No. AC167730250
Change view
Click to view available options
25L; Metal drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC167730250 25L; Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC167730250 Supplier Thermo Scientific Chemicals Supplier No. 167730250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For UV/VIS spectroscopy

Specifications

Absorbance 0.005 max. at 280nm, 0.01 max. at 270nm, 0.075 max. at 260nm, 0.5 max. at 250nm, 1 max. at 245nm
CAS 67-66-3, 513-35-9
Density 1.4800g/mL
Quantity 25L
Packaging Metal Drum
Residue after Evaporation 0.0003% max.
Specific Gravity 1.48
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
MDL Number MFCD00000826
Water 0.03% max.
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Chloroform, Stabilized with amylene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only