missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, 99.9% min., OmniSolv™, MilliporeSigma™
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Assay Percent Range | 99.9% min. |
| Formula Weight | 119.38 |
| Chemical Name or Material | Chloroform |