missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99.8% min., ACS, MilliporeSigma™

Catalog No. MCX10556
Change view
Click to view available options
Quantity:
500 mL
1 L
295 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX10556 500 mL
MCX10552 1 L
MCX105520 295 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MCX10556 Supplier Technidata Supplier No. CX10556

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.8% min.
Formula Weight 119.38
Chemical Name or Material Chloroform, For ACS analysis