missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ≥99.5%, Solstice

Catalog No. 60047879
Change view
Click to view available options
Quantity:
4 x 25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047879 4 x 25 mL Glass bottle
60047881 500 mL Glass bottle
60047876 1 L Glass bottle
60047877 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60047879 Supplier Solstice Supplier No. C24324X25ML

Contains 100-200 ppm amylenes as stabilizer,≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -61.3°C
Color Colorless
Density 1.492g/mL at 25°C
Boiling Point 61°C (1013 hPa)
Odor Sweet
Packaging Glass bottle
Quantity 4 x 25 mL
UN Number UN1888
Beilstein 1731042
IUPAC Name trichloromethane
Stabilizer Amylenes
Vapor Density 4.1 (vs air)
Vapor Pressure 160mmHg (20°C)
Formula Weight 119.38g/mol
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Chloroform
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338-P311
DOTInformation 6.1/PGIII
RTECSNumber 200-663-8