missing translation for 'onlineSavingsMsg'
Learn More

Chloroethane (ca. 15% in Tetrahydrofuran, ca. 2.0mol/L), TCI America™

Catalog No. C2883100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2883100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2883100ML Supplier TCI America Supplier No. C2883100ML
Only null left

Chemical Identifiers

CAS 75-00-3
Molecular Formula C2H5Cl
Molecular Weight (g/mol) 64.512
MDL Number MFCD00000961
InChI Key HRYZWHHZPQKTII-UHFFFAOYSA-N
Synonym ethyl chloride, ethane, chloro, chlorethyl, monochloroethane, chlorene, chelen, muriatic ether, monochlorethane, anodynon, cloretilo
PubChem CID 6337
ChEBI CHEBI:47554
IUPAC Name chloroethane
SMILES CCCl

Specifications

Color Colorless
Quantity 100 mL
UN Number 1993
Formula Weight 64.51
Physical Form Liquid
Chemical Name or Material Chloroethane (ca. 15% in Tetrahydrofuran, ca. 2.0mol/L)
RTECSNumber KH7525000
TSCA Yes
Recommended Storage Refrigerator