missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylsilane, tech. 90%

Catalog No. p-7043944
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0360206 5 g
AAL0360214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0360206 Supplier Thermo Scientific Chemicals Supplier No. L0360206
Only null left

CAS: 1631-83-0 | C12H10ClSi | 217.747 g/mol

Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.

Chemical Identifiers

CAS 1631-83-0
Molecular Formula C12H10ClSi
Molecular Weight (g/mol) 217.747
MDL Number MFCD00045144
InChI Key YCITZMJNBYYMJO-UHFFFAOYSA-N
Synonym chlorodiphenylsilane, diphenylchlorosilane, chloro diphenyl silane, silane, chlorodiphenyl, benzene, 1,1'-chlorosilylene bis, c12h11clsi, chlorodiphenylsilyl, diphenylsilyl chloride, chloro diphenyl silicon
PubChem CID 6327323
IUPAC Name chloro(diphenyl)silicon
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)Cl

Specifications

Boiling Point 280°C
Quantity 5 g
UN Number UN2987
Beilstein 2937164
Flash Point 105°C (221°F)
Solubility Information Reacts with water.
Formula Weight 218.76
Percent Purity 90%
Grade Technical
Refractive Index 1.579
Sensitivity Moisture sensitive
Density 1.123
Chemical Name or Material Chlorodiphenylsilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 216-634-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only