missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodiphenylsilane, tech. 90%
Description
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
Specifications
Specifications
| Boiling Point | 280°C |
| Quantity | 25 g |
| UN Number | UN2987 |
| Beilstein | 2937164 |
| Flash Point | 105°C (221°F) |
| Solubility Information | Reacts with water. |
| Formula Weight | 218.76 |
| Percent Purity | 90% |
| Grade | Technical |
| Refractive Index | 1.579 |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| EINECSNumber | 216-634-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only