missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylphenylsilane 96.0+%, TCI America™

Catalog No. D11475ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11475ML 5 mL
D114725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11475ML Supplier TCI America Supplier No. D11475ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chlorodimethylphenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 5 mL
UN Number 2987
Formula Weight 170.71
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethylphenylsilane
EINECSNumber (3)-2641
TSCA Yes