missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylphenylsilane, 97+%

CAS: 768-33-2 | C8H11ClSi | 170.71 g/mol

Supplier:  Thermo Scientific Chemicals A1563814

 View more versions of this product

Catalog No. AAA1563814


Only null left
Explore available promotions

Description

Description

Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

Solubility
Miscible with tetrahydrofuran, ether and hexane.

Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
Specifications

Specifications

192°C to 193°C
768-33-2
C8H11ClSi
UN2987
72°C (161°F)
Miscible with tetrahydrofuran,ether and hexane.
C[Si](C)(Cl)C1=CC=CC=C1
170.71
170.72
1.509
1.032
Chlorodimethylphenylsilane
25 g
MFCD00000499
606292
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
KWYZNESIGBQHJK-UHFFFAOYSA-N
chloro-dimethyl-phenylsilane
13029
≥97%
Moisture sensitive

Chemical Identifiers

768-33-2
170.71
KWYZNESIGBQHJK-UHFFFAOYSA-N
13029
C[Si](C)(Cl)C1=CC=CC=C1
C8H11ClSi
MFCD00000499
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
chloro-dimethyl-phenylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

EINECSNumber : 212-193-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only