missing translation for 'onlineSavingsMsg'
Learn More

chlorodimethylphenylsilane, 95%

CAS: 768-33-2 | C8H11ClSi | 170.71 g/mol

$237.01 - $237.01

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chloro-dimethyl-phenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151070250
SDS View Documents
Thermo Scientific Chemicals
151070250
25 g Glass bottle
Each for $237.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

768-33-2
170.71
KWYZNESIGBQHJK-UHFFFAOYSA-N
13029
C[Si](C)(Cl)C1=CC=CC=C1
C8H11ClSi
MFCD00000499
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
chloro-dimethyl-phenylsilane

Specifications

768-33-2
1.0320g/mL
61°C
94% min. (GC)
C8H11ClSi
C6H5Si(CH3)2Cl
25 g
1.032
Solubility in water: may decompose
C[Si](C)(Cl)C1=CC=CC=C1
170.71
170.72
Liquid
Colorless to Yellow
80°C to 84°C (16.0 mmHg)
Authentic
Glass bottle
1.5060 to 1.51
MFCD00000499
02,278
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
KWYZNESIGBQHJK-UHFFFAOYSA-N
chloro-dimethyl-phenylsilane
13029
95%
Chlorodimethylphenylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 212-193-

SDS
Documents

Documents

RUO – Research Use Only