Learn More
Chlorodimethyl(pentafluorophenyl)silane, 96%
CAS: 20082-71-7 | C8H6ClF5Si | 260.66 g/mol
Supplier: Thermo Scientific Chemicals L0055003
Description
It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| 78°C to 81°C (10mmHg) | |
| 20082-71-7 | |
| C8H6ClF5Si | |
| UN2987 | |
| 95°C (203°F) | |
| Reacts with water. | |
| C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 260.66 | |
| 260.66 | |
| 1.447 | |
| 1.381 |
| Chlorodimethyl(pentafluorophenyl)silane | |
| 1 g | |
| MFCD00000500 | |
| 2858407 | |
| pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane | |
| PQRFRTCWNCVQHI-UHFFFAOYSA-N | |
| chloro-dimethyl-(2,3,4,5,6-pentafluorophenyl)silane | |
| 88361 | |
| 96% | |
| Moisture sensitive |
Chemical Identifiers
| 20082-71-7 | |
| 260.66 | |
| PQRFRTCWNCVQHI-UHFFFAOYSA-N | |
| 88361 | |
| C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F |
| C8H6ClF5Si | |
| MFCD00000500 | |
| pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane | |
| chloro-dimethyl-(2,3,4,5,6-pentafluorophenyl)silane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber : 243-506-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only