missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl(3-phenylpropyl)silane 97.0+%, TCI America™

Catalog No. p-7125011
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14885ML 5 mL
C148825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14885ML Supplier TCI America Supplier No. C14885ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17146-09-7
Molecular Formula C11H17ClSi
Molecular Weight (g/mol) 212.79
MDL Number MFCD00049281
InChI Key ASSMBLOISZSMMP-UHFFFAOYSA-N
PubChem CID 86971
IUPAC Name chlorodimethyl(3-phenylpropyl)silane
SMILES C[Si](C)(Cl)CCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 mL
UN Number 2987
Formula Weight 212.79
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethyl(3-phenylpropyl)silane