missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiisopropylphosphine, 96%, AcroSeal™

Catalog No. AC427270500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427270500 50 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427270500 Supplier Thermo Scientific Chemicals Supplier No. 427270500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40244-90-4 | C6H14ClP | 152.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40244-90-4
Molecular Formula C6H14ClP
Molecular Weight (g/mol) 152.61
MDL Number MFCD00015027
InChI Key JZPDBTOWHLZQFC-UHFFFAOYSA-N
Synonym chlorodiisopropylphosphine, diisopropylchlorophosphine, di-i-propylchlorophosphine, phosphinous chloride, bis 1-methylethyl, ipr2pcl, pubchem6476, chlorodiisopropylphosphane, chloro diisopropylphosphine, acmc-209jc6, diisopropylphosphinyl chloride
PubChem CID 538967
IUPAC Name chloro-di(propan-2-yl)phosphane
SMILES CC(C)P(C(C)C)Cl

Specifications

Color Yellow
Density 0.9500g/mL
Boiling Point 69.0°C (33.0 mmHg)
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4740 to 1.4760
Linear Formula [(CH3)2CH]2PCl
Quantity 50 mL
Specific Gravity 0.95
Formula Weight 152.61
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Chlorodiisopropylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only