missing translation for 'onlineSavingsMsg'
Learn More

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium, 97%

Catalog No. p-4258892
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435122500 250 mg Glass bottle
AC435120010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435122500 Supplier Thermo Scientific Chemicals Supplier No. 435122500
Only null left

CAS: 791629-96-4 | C16H37NP2·ClH2Ir | 535.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 791629-96-4
Molecular Formula C16H37NP2·ClH2Ir
Molecular Weight (g/mol) 535.1
InChI Key QPAONQONOKDMMJ-UHFFFAOYSA-M
Synonym iridium 3+ bis 2-diisopropylphosphanyl ethyl amine chloride dihydride, iridium 3+ chloride hydride-2-di propan-2-yl phosphanyl-n-2-di propan-2-yl phosphanyl ethyl ethan-1-amine 1/1/2/1

Specifications

Melting Point 240.6°C to 241.9°C
Color Tan to White
Elemental Analysis 34.50 to 37.30% C, 2.50 to 2.70% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 535.1
Percent Purity 97%
Physical Form Powder
Chemical Name or Material chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only