missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclopentane 98.0+%, TCI America™

Catalog No. p-7124501
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C056625ML 25 mL
C0566500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C056625ML Supplier TCI America Supplier No. C056625ML

Chemical Identifiers

CAS 930-28-9
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00001360
InChI Key NDTCXABJQNJPCF-UHFFFAOYSA-N
Synonym cyclopentane, chloro, cyclopentyl chloride, chloroclopentane, cyclopentylchloride, unii-to3f41cl9a, 1-chlorocyclopentane, to3f41cl9a, cyclopentylchlorid, chloro-cyclopentan, chlorocyclopenetane
PubChem CID 70252
IUPAC Name chlorocyclopentane
SMILES C1CCC(C1)Cl

Specifications

Color Yellow
Boiling Point 114°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorocyclopentane
EINECSNumber (3)-4323
TSCA Yes