missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclohexane 98.0+%, TCI America™

Catalog No. C0153500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0153500ML 500 mL
C015325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0153500ML Supplier TCI America Supplier No. C0153500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 542-18-7
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00003822
InChI Key UNFUYWDGSFDHCW-UHFFFAOYSA-N
Synonym cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
PubChem CID 10952
ChEBI CHEBI:39156
IUPAC Name chlorocyclohexane
SMILES C1CCC(CC1)Cl

Specifications

Melting Point -44°C
Color Colorless
Boiling Point 143°C
Quantity 500 mL
UN Number 1993
Formula Weight 118.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorocyclohexane
EINECSNumber (3)-2252
RTECSNumber GU8685000
TSCA Yes