missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorocyclobutane 98.0+%, TCI America™
$141.67 - $512.62
Chemical Identifiers
| CAS | 1120-57-6 |
|---|---|
| Molecular Formula | C4H7Cl |
| Molecular Weight (g/mol) | 90.55 |
| MDL Number | MFCD00013736 |
| InChI Key | STJYMUBZVMSMBP-UHFFFAOYSA-N |
| Synonym | cyclobutyl chloride, cyclobutane, chloro, chloro-cyclobutane, cyclobutylchloride, chloranylcyclobutane, 1-chlorocyclobutane, acmc-1butg, chlorocyclobutane, 97+% |
| PubChem CID | 70712 |
| IUPAC Name | chlorocyclobutane |
| SMILES | C1CC(C1)Cl |
Chemical Identifiers
| 1120-57-6 | |
| 90.55 | |
| STJYMUBZVMSMBP-UHFFFAOYSA-N | |
| 70712 | |
| C1CC(C1)Cl |
| C4H7Cl | |
| MFCD00013736 | |
| cyclobutyl chloride, cyclobutane, chloro, chloro-cyclobutane, cyclobutylchloride, chloranylcyclobutane, 1-chlorocyclobutane, acmc-1butg, chlorocyclobutane, 97+% | |
| chlorocyclobutane |
Specifications
| 1120-57-6 | |
| 82°C | |
| MFCD00013736 | |
| 1993 | |
| STJYMUBZVMSMBP-UHFFFAOYSA-N | |
| chlorocyclobutane | |
| 70712 | |
| ≥98.0% (GC) | |
| Chlorocyclobutane |
| Colorless | |
| C4H7Cl | |
| 1 mL | |
| cyclobutyl chloride, cyclobutane, chloro, chloro-cyclobutane, cyclobutylchloride, chloranylcyclobutane, 1-chlorocyclobutane, acmc-1butg, chlorocyclobutane, 97+% | |
| C1CC(C1)Cl | |
| 90.55 | |
| 90.55 | |
| Liquid |
Safety and Handling
TSCA : No