missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclobutane 98.0+%, TCI America™

Catalog No. C10335ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10335ML 5 mL
C10331ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10335ML Supplier TCI America Supplier No. C10335ML

Chemical Identifiers

CAS 1120-57-6
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00013736
InChI Key STJYMUBZVMSMBP-UHFFFAOYSA-N
Synonym cyclobutyl chloride, cyclobutane, chloro, chloro-cyclobutane, cyclobutylchloride, chloranylcyclobutane, 1-chlorocyclobutane, acmc-1butg, chlorocyclobutane, 97+%
PubChem CID 70712
IUPAC Name chlorocyclobutane
SMILES C1CC(C1)Cl

Specifications

Color Colorless
Boiling Point 82°C
Quantity 5 mL
UN Number 1993
Formula Weight 90.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorocyclobutane