missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorocholine Chloride 98.0+%, TCI America™
$115.51 - $962.59
Chemical Identifiers
| CAS | 999-81-5 |
|---|---|
| Molecular Formula | C5H13Cl2N |
| Molecular Weight (g/mol) | 158.066 |
| MDL Number | MFCD00011869 |
| InChI Key | UHZZMRAGKVHANO-UHFFFAOYSA-M |
| Synonym | chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan |
| PubChem CID | 13836 |
| IUPAC Name | 2-chloroethyl(trimethyl)azanium;chloride |
| SMILES | C[N+](C)(C)CCCl.[Cl-] |
Chemical Identifiers
| 999-81-5 | |
| 158.066 | |
| UHZZMRAGKVHANO-UHFFFAOYSA-M | |
| 13836 | |
| C[N+](C)(C)CCCl.[Cl-] |
| C5H13Cl2N | |
| MFCD00011869 | |
| chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
| 2-chloroethyl(trimethyl)azanium;chloride |
Specifications
| 999-81-5 | |
| C5H13Cl2N | |
| 25 g | |
| chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
| C[N+](C)(C)CCCl.[Cl-] | |
| 158.066 | |
| 158.07 | |
| Crystalline Powder |
| White | |
| MFCD00011869 | |
| 2811 | |
| UHZZMRAGKVHANO-UHFFFAOYSA-M | |
| 2-chloroethyl(trimethyl)azanium;chloride | |
| 13836 | |
| ≥98.0% (T) | |
| Chlorocholine Chloride |
Safety and Handling
RTECSNumber : BP5250000
TSCA : Yes