missing translation for 'onlineSavingsMsg'
Learn More

Chlorure de chlorocholine 98,0+%, TCI America™

Numéro de catalogue. C0172500G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C0172500G 500 g
C017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C0172500G Fournisseur TCI America Code fournisseur C0172500G

Identifiants chimiques

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroéthyl(triméthyl)azanium; Chlorure
SMILES C[N+](C)(C)CCCl.[Cl-]

Spécifications

Color Blanc
Quantity 500 g
UN Number 2811
Formula Weight 158.07
Percent Purity ≥98.0% (T)
Physical Form Poudre cristalline
Chemical Name or Material Chlorocholine Chloride
RTECSNumber BP5250000
TSCA Yes