missing translation for 'onlineSavingsMsg'
Learn More

Chlorocholine Chloride 98.0+%, TCI America™

Catalog No. C017225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C017225G 25 g
C0172500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C017225G Supplier TCI America Supplier No. C017225G

Chemical Identifiers

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCCl.[Cl-]

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 158.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorocholine Chloride
RTECSNumber BP5250000
TSCA Yes