missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(cyclooctene)rhodium(I) Dimer 96.0+%, TCI America™

Catalog No. C2253500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2253500MG 500 mg
C2253100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2253500MG Supplier TCI America Supplier No. C2253500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12279-09-3
Molecular Formula C32H56Cl2Rh2
Molecular Weight (g/mol) 717.51
MDL Number MFCD00013287
InChI Key GQPAPAIPOLEZHT-XFCUKONHSA-L
Synonym chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer
PubChem CID 53384308
IUPAC Name bis(λ¹-rhodium(1+)) tetrakis((Z)-cyclooctene) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1

Specifications

Melting Point 190°C
Quantity 500 mg
Formula Weight 717.51
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorobis(cyclooctene)rhodium(I) Dimer
TSCA No
Recommended Storage Freezer