missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(cyclooctene)rhodium(I) dimer, 98%

Catalog No. p-4813401
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369371000 100 mg Glass bottle
AC369375000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC369371000 Supplier Thermo Scientific Chemicals Supplier No. 369371000
Only null left

CAS: 12279-09-3 | C32H56Cl2Rh2 | 717.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12279-09-3
Molecular Formula C32H56Cl2Rh2
Molecular Weight (g/mol) 717.51
MDL Number MFCD00013287
InChI Key GQPAPAIPOLEZHT-XFCUKONHSA-L
Synonym chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer
PubChem CID 53384308
IUPAC Name cyclooctene;rhodium;dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1

Specifications

Melting Point 187.0°C to 191.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 717.52
Percent Purity 98%
Chemical Name or Material Chlorobis(cyclooctene)rhodium(I) dimer
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only