missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, Spectrophotometric Grade, 99.9%

Catalog No. p-4824673
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22921AP 500 mL
AA22921K2 1 L
AA22921M1 4 x 1 L
AA22921K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA22921AP Supplier Thermo Scientific Chemicals Supplier No. 022921AP
Only null left

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

Chlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point °C to 45°C
Color Colorless
Appearance Colorless liquid
Density 1.106
Boiling Point 132°C
Quantity 500 mL
Flash Point 28°C (82°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.524
Assay Percent Range 99.9%
Vapor Pressure 12 hPa (9mm Hg) at 20°C
Beilstein 605632
Solubility Information Miscible with methanol and diethyl ether. Immiscible with water.
Formula Weight 112.56
Grade Spectrophotometric
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Category H226-H315-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: III
EINECSNumber 203-628-5
RTECSNumber CZ0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only