missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ReagentPlus™, 99%, Honeywell™

Catalog No. p-7120315
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044637 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044637 Supplier Honeywell Chemicals Supplier No. 1013892.5L
Only null left

Reagent Grade, 99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Color Colorless
Density 1.106g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.071
Flash Point 27°C (80.6°F)
Ignition Point 1178°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.524(lit.)
Vapor Pressure 11.8mmHg (25°C)
UN Number UN1134
Beilstein 605632
Boiling Point 130°C to 132°C
Solubility Information Soluble in water, 0.207g/L at 20°C
Formula Weight 112.56g/mol
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Statement P210-P261-P370 + P378
Precautionary Statement H226-H315-H332-H411
DOTInformation class 3 - PG 3 - Chlorobenzene
EINECSNumber 203-628-5
RTECSNumber CZ0175000