missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene (Laboratory), Fisher Chemical™
Specifications
Specifications
| Melting Point | -45°C |
| Density | 1.108g/cm³ |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 12mbar at 20°C |
| Viscosity | 0.8 mPa/s at 20°C |
| UN Number | 1134 |
| Formula Weight | 112.56 |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:3 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |